Learn More
Triforest Enterprises 250ML sterile PETG square media bottle with standard cap and heat-shrink band around closure and neck for tamper-evident seal
Supplier:  Triforest Enterprises BGC0250SB
Encompass_Preferred
TriForest Square PETG media bottles are ideal for storage of sterile reagents. Bottles are pre-sterilized, SAL 10-6, and include a standard cap with heat-shrink band around closure and neck for tamper-evident seal. Each tray is packaged for sterile production with multi-barrier packaging.
Catalog No. 50-607-666
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.