Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diphenylphosphine

Catalog No. 25296450G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-296-450G 50 g
25-296-41KG 1 kg
25-296-410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 25-296-450G Supplier Sigma Aldrich Supplier No. 25296450G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Deprotonation of Ph2PH gives diphenylphosphide, used for synthesis of novel phosphine ligands, Wittig-Horner reagents, and phosphonium salts; source of phosphorus-centered radicals.

Specifications

CAS 829-85-6
Density 1.07 g/mL at 25°C
Boiling Point 280°C
Molecular Formula C12H11P
Linear Formula (C6H5)2PH
Refractive Index n20/D 1.625
MDL Number MFCD00003040
Quantity 50 g
Synonym HPPh2; Ph2PH
Molecular Weight (g/mol) 186.19 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.