Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tripropyl phosphate

Catalog No. 2553275G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-532-75G 5 g
25-532-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-532-75G Supplier Sigma Aldrich Supplier No. 2553275G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tripropyl phosphate is an organophosphorus flame retardant and plasticizer.Tripropyl phosphate has been used as external standard in sampling of organophosphate triesters in indoor air by using solid phase extraction cartridges.

Specifications

CAS 513-08-6
Density 1.012 g/mL (at 25°C (literature))
Boiling Point 120°C to 122°C (10 mmHg)
Molecular Formula C9H21O4P
Linear Formula (CH3CH2CH2O)3 P(O)
Refractive Index n20/D 1.416 (literature)
Quantity 5 g
MDL Number MFCD00009368
Molecular Weight (g/mol) 224.23
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.