Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,4'-Methylenebis(phenyl isocyanate)

Catalog No. 256439500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
500 g
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-643-9500G 500 g
25-643-95KG 5 kg
25-643-950G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 25-643-9500G Supplier Sigma Aldrich Supplier No. 256439500G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,4'-Methylenebis(phenyl isocyanate)

Specifications

CAS 101-68-8
Melting Point 42°C to 45°C
Density 1.18 g/mL (at 25°C)
Boiling Point 200°C (5 mmHg)
Molecular Formula C15H10N2O2
Linear Formula CH2(C6H4NCO)2
Quantity 500 g
MDL Number MFCD00036131
Synonym 4,4 -MDI; Bis(4-isocyanatophenyl)methane
Molecular Weight (g/mol) 250.25
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.