Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Protoporphyrin IX disodium salt

Catalog No. 2583851G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-838-51G 1 g
25-838-55G 5 g
25-838-5250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 25-838-51G Supplier Sigma Aldrich Supplier No. 2583851G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Protoporphyrin disodium salt (PpIX) was used as a dye content for fluorescence spectroscopic characterization in a study. The cytotoxic effect of PpIXon sarcoma 180 solid tumors cells was reported in a study.

Specifications

CAS 50865-01-5
Molecular Formula C34H32N4Na2O4
Linear Formula C34H32N4Na2O4
Quantity 1 g
MDL Number MFCD00151108
Synonym 3,7,12,17-Tetramethyl-8,13-divinyl-2,18-porphinedipropionic acid disodium salt; Depocolin-S; Ooporphyrin disodium salt
Molecular Weight (g/mol) 606.62
Percent Purity ≥90% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.