Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzoyl Chloride

Catalog No. 2599501L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-995-01L 1 L
25-995-0100ML 100 mL
25-995-0250ML 250 mL
25-995-05ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 25-995-01L Supplier Sigma Aldrich Supplier No. 2599501L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzoyl Chloride

Specifications

CAS 98-88-4
Melting Point -1°C
Density 1.211 g/mL at 25°C
Boiling Point 198°C
Molecular Formula C7H5ClO
Refractive Index n20/D 1.553
Linear Formula C6H5COCl
MDL Number MFCD00000653
Quantity 1 L
Molecular Weight (g/mol) 140.57 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.