Learn More
Cayman Chemical Myriocin, 35891-70-4, 25 mg
SDP
Supplier:  Cayman Chemical 6315025
Encompass_Preferred
Myriocin is an amino fatty acid antibiotic derived from certain thermophylic fungi, in this case Mycelia sterilia. It is a potent immunosuppressant having 10- to 100-fold more activity than cyclosporin A.{8482} Myriocin is a potent inhibitor of serine palmitoyltransferase (Ki = 0.28 nM), the enzyme that catalyzes the first step of sphingolipid biosynthesis. {14764} It disrupts substratum adhesion of melanoma cells.{14763} It also suppresses cell proliferation in the murine cytotoxic T cell line CTLL-2 (IC50 = 15 nM) via apoptosis.{14764,2816} Myriocin suppresses replication of the hepatitis C virus (HCV) in a murine model.{14762}
Catalog No. NC1462668
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.