Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bacterial Alkaline Phosphatase

Catalog No. 18011015
Encompass_Preferred
Change view
Click to view available options
Quantity:
2500 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-011-015 2500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-011-015 Supplier Invitrogen™ Supplier No. 18011015
  • $405.00
    $384.65 / Each of 1

    Save $20.35

    5% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

A vial of 10X dephosphorylation buffer [100mM Tris-HCl (pH 8.0)].

Removes 3' and 5' phosphates from DNA and RNA

BAP is active at 65°C for at least 1 hr. and is inactivated by phenol extraction

  • Source: Purified from E. coli C90
  • Performance and Quality Testing: Endodeoxyribonuclease, exodeoxyribonuclease, and ribonuclease assays; dephosphorylation efficiency determined
  • Unit Definition: One unit hydrolyzes 1nmol of ATP in 30 min. at 37°C
  • Unit Reaction Conditions: 10mM Tris-HCl (pH 8.0), 1.5mM [γ-32P]ATP, and enzyme in 50μL for 30 min. at 37°C

Dephosphorylation of 5'-phosphorylated termini of vector DNA to prevent self-ligation; Dephosphorylation of 5' termini of nucleic acids prior to forward reaction with kinase; Cloning; Restriction Enzyme Cloning

Order Info

Shipping Conditions: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Bacterial Alkaline Phosphatase is supplied with a vial of 10X dephosphorylation buffer [100 mM Tris-HCl (pH 8.0)]. Store at -20°C.
Shipping Condition Wet Ice
Enzyme BAP
Quantity 2500 U
Product Type Alkaline Phosphatase
Why are alkaline phosphatases used in cloning protocols? What is a typical protocol for dephosphorylation of nucleic acids?

Alkaline phosphatases are used to dephosphorylate the 5' ends of DNA. In cloning, it is used to prevent self-ligation of vector DNA. Standard ligation of DNA with ligase requires a 5' phosphate to be present on at least one of the ends being joined. When a DNA insert containing phosphates on both 5' termini is added to a dephosphorylated vector, the insert will be efficiently ligated into the vector, but the vector will not be able to self-ligate. Thus, dephosphorylation of vector lowers the number of background colonies containing vector without insert.

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.