Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyceryl Monostearate, Powder, Food Grade, Spectrum™ Chemical
SDP

Catalog No. 18603003 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-003 45 kg Poly Pail
18-603-000 500 g Poly Pail
18-603-001 2.5 kg Poly Pail
18-603-002 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-003 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. GL14945KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GL149, 31566-31-1, C21H42O4

Spectrum™ Chemical Glyceryl Monostearate, Powder, Food Grade is commonly referred to as GMS and is an organic molecule that is often used as an emulsifier. It has neither color nor odor but has a sweet taste and is a flaky hygroscopic powder. It is naturally occurring in the body and is the by product of fat breakdowns. In addition to use as an emulsifier, it is also an anti-caking and thickening agent.

Chemical Identifiers

CAS 31566-31-1
Molecular Formula C21H42O4
Molecular Weight (g/mol) 358.56
InChI Key VBICKXHEKHSIBG-UHFFFAOYNA-N
IUPAC Name 2,3-dihydroxypropyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO

Specifications

CAS Max % 100%
Packaging Poly Pail
Quantity 45 kg
Percent Purity 100%
Grade Food
Physical Form Crystals or Flakes
Chemical Name or Material Glyceryl Monostearate, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.