Learn More
Chemglass Life Sciences Chemglass Pyrex Erlenmeyer Flask, Narrow Mouth, 25 mL, Heavy Duty Rim, Graduated
Supplier: Chemglass Life Sciences CG849625
Narrow-mouth 25 mL Erlenmeyer flask made from heat-resistant Pyrex glass for general laboratory mixing, heating, and storage. The heavy-duty rim and durable white-enamel graduations provide robustness and easy measurement while an extra-large marking area supports sample identification.
- 25 mL capacity.
- Pyrex glass construction.
- Narrow mouth for controlled pouring and reduced evaporation.
- Heavy-duty rim for added durability.
- Approximate graduations in durable white enamel.
- Extra-large marking space.
- Height 65 mm; widest diameter 41 mm.
- Stopper size 0.
- Not intended for vacuum use.