Learn More
Thomas Scientific 25mL Pipet, Individually Wrapped Packed in Bags, Sterile
SDP
Supplier:  Thomas Scientific 1222B04
Encompass
NEW – Now available in 100mL size for large volume pipetting Peel-to-open plastic/plastic wrappers are the ideal option for eliminating particulate; proven benefit over traditional paper/plastic wrapper Wrappers reduce static cling versus other brands of plastic wrap pipets All plastic wrapper can be wiped with alcohol without smearing ink or penetrating paper Color coded stripes and packaging enable quick volume identification Bi-directional graduations are printed in bold easy-to-read black ink Premium material filter plug
Catalog No. NC0635733
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.