Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Accessories for Glass Microanalysis Filter Holders

Catalog No. XX1012503 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Base with Stopper, Screen and Gasket
Base, Stopper and Screen
Fritted Glass Base with Stopper
Glass Funnel
Perforated Stopper
Spring Clamp
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
XX1012503 Spring Clamp
XX1012508 Perforated Stopper
XX1012542 Base with Stopper, Screen and Gasket
XX1012532 Base, Stopper and Screen
XX1012514 Glass Funnel
XX1012502 Fritted Glass Base with Stopper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. XX1012503 Supplier MilliporeSigma™ Supplier No. XX1012503
Add to Cart
Add to Cart

Accessories for filter holders for sample preparation.

Millipore Glass Vacuum Filtration Accessory Product:

  • These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
  • This item is an accessory part for the Millipore 25 mm Glass Microanalysis Filter Holder Kits.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Disposable Reusable
For Use With (Equipment) 25 mm Glass Microanalysis Vacuum Filter
Material Anodized Aluminum
Sterility Non-sterile
Type Spring Clamp
Diameter (English) Inside 0.98 in.
Diameter (Metric) Inside 25 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.