Learn More
Delta Technologies Ltd Unpolished Float Glass, 25 x 75 x 1.1 mm, SiO2 passivated / Indium Tin Oxide coated one surface, Rs = 70 - 100 ohms, cut edges., Qty 100
SDP
Supplier:  Delta Technologies Ltd CG90INS115
Encompass
ITO Coated Slides, Unpolished Float Glass, 25 x 75 x 1.1 mm, SiO2 passivated / Indium Tin Oxide coated one surface, resistance = 70 - 100 ohms/sq. Delta Technologies, Limited offers a variety of transparent, conductive indium tin oxide (ITO), flourine-doped tin oxide (FTO) coated, and uncoated products on several different substrates, custom cut to any dimension required.
Catalog No. NC1365838
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.