Learn More
Biotium H2DCFDA (2′,7′-Dichlorodihydrofluorescein diacetate)
Supplier:  Biotium 10058
Encompass_Preferred
H2DCFDA (2′,7′-Dichlorodihydrofluorescein diacetate) is a useful fluorogenic reagent to detect reactive oxygen intermediates in cells. On oxidation, H2DCFDA becomes the highly green fluorescent 2′,7′-dichlorofluorescein. Properties: Ex/Em nm (pH >6) = 504/529 nm (end product); White solid soluble in DMSO or DMF; Store desiccated at -20°C and protect from light; C24H16Cl2O7; MW: 487.29; CAS No. [4091-99-0]
Catalog No. NC0667146