Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences Dish, Crystallizing, 270mL, 90 x 50mm
SDP

Catalog No. NC0563003
Encompass_Preferred

Dish, Crystallizing, 270mL, 90 x 50mm Old Part Number MW-108-03

Catalog No. NC0563003 Supplier Chemglass Life Sciences Supplier No. CG827690
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dish, Crystallizing, 270mL, 90 x 50mm Old Part Number MW-108-03

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.