Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butylpyrrolidine

Catalog No. 280372100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-037-2100ML 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28-037-2100ML Supplier Sigma Aldrich Supplier No. 280372100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butylpyrrolidine

Specifications

CAS 767-10-2
Boiling Point 155°C to 157°C (754 mmHg)
Molecular Formula C8H17N
Refractive Index n20/D 1.44 (literature)
Linear Formula C8H17N
MDL Number MFCD00003184
Density 0.814 g/mL (at 25°C (literature))
Quantity 25 mg
Molecular Weight (g/mol) 127.23
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.