Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Amyl methyl ether

Catalog No. 283096100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-309-6100ML 100 mL
28-309-6500ML 500 mL
28-309-65ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 28-309-6100ML Supplier Sigma Aldrich Supplier No. 283096100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Tert-amyl methyl ether is an fuel ether oxygenate and has been quantitated in water by membrane extraction ion mobility spectrometry
  • Tert-amyl methyl ether is also referred to as 2-methoxy-2-methyl butane
  • Kinetics of ion exchange resin (amberlyst 18) catalyzed tert-amyl methyl ether reactions have been investigated

Specifications

CAS 994-05-8
Density 0.76-0.78 g/mL (at 25°C (literature))
Boiling Point 85°C to 86°C (lit.)
Molecular Formula C6H14O
Refractive Index n20/D 1.389 (literature); n20/D 1.3896
Linear Formula C2H5C(CH3)2 OCH3
MDL Number MFCD00010186
Quantity 100 mL
Synonym 1,1-Dimethylpropyl methyl ether; Methyl 2-methyl-2-butyl ether; Methyl tert-pentyl ether; TAME
Molecular Weight (g/mol) 102.17
Percent Purity 97%
Biological Activity Central Nervous System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.