Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Trifluoromethyl)benzyl bromide

Catalog No. 2905565G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-055-65G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 29-055-65G Supplier Sigma Aldrich Supplier No. 2905565G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(Trifluoromethyl)benzyl bromide

Specifications

CAS 402-23-3
Density 1.565 g/mL (at 25°C (literature))
Boiling Point 69°C (4 mmHg)
Molecular Formula C8H6BrF3
Refractive Index n20/D 1.492 (literature)
Linear Formula CF3C6H4CH2Br
MDL Number MFCD00000395
Quantity 100 mg
Synonym alpha-Bromo-alpha,alpha,alpha-trifluoro-m-xylene
Molecular Weight (g/mol) 239.03
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.