Learn More
Expression Systems 293-ES cells, Suspension in ESF SFM, 20M total cells in 15mL conical tube
SDP
Supplier:  Expression Systems 94007S
Encompass_Preferred
Expression Systems offers 293 cell lines adapted to serum-free suspension culture in ESF SFM. The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
293-ES cells are derived from the human embryonic kidney HEK 293 cell line. 293-ES cells are adapted for suspension culture in ESF SFM and are available as a frozen vial or suspension culture.
Capable of producing high levels of AAV vectors
Capable of high levels of recombinant protein expression
Good growth in adherent or suspension culture Quality and performance testing
Organism: Homo sapiens, human
Tissue: Embryonic Kidney
Cell Type: Epithelial
Biosafety Level 2: Cells contain adenovirus. Biosafety classification is based on U.S. Public Health Service Guidelines, it is the responsibility of the customer to ensure that their facilities comply with biosafety regulations for their own country.
Catalog No. NC1440234
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.