Learn More
Specifications
Specifications
| Cell Line | 293T |
| Description | Whole cell lysate (denatured) |
| Lysis Buffer | 1X Sample Buffer (0.2 M Tris-HCl, 2% SDS, 10% glycerol, 0.2% 2-mercaptoethanol, 0.2% Bromophenol blue). |
| Preparation Method | Culture cells in 6-well plate to 80% confluent, remove media and then rinse with cold 1x PBS. Scrape cells off the plate surface and pipette cells into microcentrifuge tube to spin down the cells. Remove PBS and then lysis and resuspend cells with 205 ul 1X sample buffer. Mix well and heat at 90°C for 10 minutes, and then cooled rapidly. |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 100 μL |
| Storage Buffer | In ready-to-use 1X Sample Buffer (0.2 M Tris-HCl, 2% SDS, 10% glycerol, 0.2% 2-mercaptoethanol, 0.2% Bromophenol blue). |
| Host Species | Human |
| Content And Storage | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| For Use With (Application) | Western Blot |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.