Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(6,6'-Dimethoxybiphenyl-2,2'-Diyl)Bis[Bis(3,5-Dimethylphenyl)Phosphine]

Catalog No. 29516100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-516-100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 29-516-100MG Supplier Sigma Aldrich Supplier No. 29516100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Atropisomeric MeOBIPHEP ligandssold in collaboration with Solvias AG

Specifications

CAS 74-98-6
Melting Point -188°C
Density 0.564 g/mL at 20°C
Boiling Point -42.1°C
Molecular Formula C3H8
Linear Formula CH3CH2CH3
Quantity 100 mg
MDL Number MFCD00009359
Molecular Weight (g/mol) 44.1 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.