Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dysprosium(III) nitrate hydrate

Catalog No. 29815825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-815-825G 25 g
29-815-8100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-815-825G Supplier Sigma Aldrich Supplier No. 29815825G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hydration may vary slightly

Specifications

CAS 100641-13-2
Molecular Formula DyN3O9 -+ xH2O
Linear Formula Dy(NO3)3-+xH2O
Quantity 25 g
MDL Number MFCD00149684
Molecular Weight (g/mol) 348.51
Percent Purity 99.9% trace metals basis
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.