Learn More
Sigma Organic Chemistry 2-Chloro-5-(tributylstannyl)thiazole | 5G | 889672-73-5 | MFCD09265501
Supplier: Sigma Organic Chemistry 7288965G
2-Chloro-5-(tributylstannyl)thiazole, 5G
About This Item:
Linear Formula: C15H28ClNSSn
Density: 1.236 g/mL at 25 C
Refractive Index: n20/D 1.528
Storage: 2-8C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.