Learn More
Chemglass Life Sciences Chemglass Cryogenic Vial, 0.5 mL, Internal Thread, Sterile, 2D Barcode, Racked
SDP
Supplier:  Chemglass Life Sciences CGN476307
Encompass_Preferred
Self-standing 0.5 mL cryogenic vial with an internal thread and permanent 2D barcode for reliable sample identification and streamlined inventory management. Sterile and manufactured from USP VI polypropylene, the vial is supplied racked in nests to facilitate handling and automated workflows.
  • Capacity: 0.5 mL
  • Thread: Internal thread
  • Barcode: Permanent 2D barcode on vial bottom
  • Sterile: E-beam sterilized (SAL = 10^-6)
  • Material: USP VI polypropylene
  • Packaging: 9 x 9 per rack; 16 racks per case (1,296 vials per case)
Catalog No. 50-273-3382