Learn More
Chemglass Life Sciences Chemglass 1.2 mL 2D-barcoded Cryogenic Vial - Self-standing, External Thread, Sterile, Racked (81/rack, 1,134/case)
SDP
Supplier:  Chemglass Life Sciences CGN476302
Encompass_Preferred
Self-standing 1.2 mL cryogenic vial with a laser-etched 2D barcode for secure sample identification and tracking. Made from USP VI polypropylene and sterilized by e-beam to SAL 10^-6, the vial is supplied racked for efficient handling and biobank compatibility.
  • 1.2 mL nominal volume.
  • External thread with special screw cap designed without silicone ring.
  • Laser-etched 2D barcode on the base; 100% inspected for readability.
  • Racked format, 9 x 9 per rack (81 vials per rack); 14 racks per case (1,134 vials per case).
  • USP VI polypropylene; non-pyrogenic; DNase/RNase and human DNA free.
  • Autoclavable; round bottom; clear graduations and white marking area.
  • Solid, one-piece construction preserves barcode integrity at low temperatures.
Catalog No. 50-273-3387