Learn More
Corning™ 2D Barcoded Cryogenic Vial Storage Box, for 100 Vials
Catalog No. 07200965 Shop All Corning Products
Change view
Click to view available options
Holds:
100 Vials
Catalog No. Holds
07-200-965 100 Vials
1 options
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 07-200-965 Supplier Corning™ Supplier No. 8674
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
For storing cryogenic vials. Corning™ 2D Barcoded Cryogenic Vial Storage Box is made of polycarbonate.
  • Reusable white polycarbonate box holds 100 cryogenic vials in a 10 x 10 array
  • Only for use with 1 - 2 mL internally threaded cryogenic vials
  • Ridges on sides of box make them easier to grip while wearing gloves
TRUSTED_SUSTAINABILITY
Specifications
Type Cryo Storage Box
Holds 100 Vials
Tube Array 10 x 10
Weight (Metric) 12 kg