Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Luer Lock insert and body connectors, injection site

Description
The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.
Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions
Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands
BPC port specifications:
• Line 1: Luer lock insert connection with 4.8 mm (3/16 in.) barb, cap, and slide clamp. Tubing: EVA + TPE 374
• Line 2: Luer lock body connection with 4.8 mm (3/16 in.) barb, cap, and slide clamp. Tubing: EVA + TPE 374
• Line 3: Tubing: EVA, injection port
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Polyethylene |
| Content And Storage | Store at room temperature. |
| Dimensions | 16 x 13.5 cm (6.3 x 5.32 in.) |
| Outer Packaging | One 4-mil. poly bag |
| Quantity | 50/Cs. |
| Sterilization Method | Irradiated |
| Shipping Condition | Room Temperature |
| Product Line | Labtainer |
| Port Detail | Luer Lock Body Connector and Injection Site |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.