Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Luer Lock insert and body connectors with reducers, injection site
GREENER_CHOICE

Catalog No. SS00030I
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SS00030I 250 mL
SS00029I 100 mL
SS00031I 500 mL
SS00032I 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SS00030I Supplier Thermo Scientific™ Supplier No. SS00030I
Add to Cart
Add to Cart

The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.

The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
Line 1: Luer lock insert connection with 4.8 mm (3/16 in.) barb, cap, slide clamp, and 4.8 x 4.1 mm (3/16 x 5/32 in.) reducer. Tubing: TPE 374, length: 30 cm (12 in.), ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Line 2: Luer lock body connection with 0.56 cm (0.22 in.) barb, plug, slide clamp, and 0.48 x 0.41 cm (0.12 x 0.16 in.) reducer. Tubing: TPE 374, length: 30 cm (12 in.), ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Line 3: Tubing: EVA, injection port

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Material Polyethylene
Content And Storage Store at room temperature.
Dimensions 16 x 13.5 cm (6.3 x 5.32 in.)
Outer Packaging One 4-mil. poly bag
Quantity 50/Cs.
Sterilization Method Irradiated
Shipping Condition Room Temperature
Product Line Labtainer
Port Detail Luer Lock Body Connector and Injection Site
Film Description ASI 26/77
Tubing Style TPE and EVA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.