Learn More
Cayman Chemical Precellys 2 mL Tissue Homogenizing Mixed Beads Kit (CKMix), 2 box
SDP
Supplier:  Cayman Chemical 104092
Encompass
Kit includes 50 preps of ceramic (zirconium oxide) mix beads of 1.4 mm and 2.8 mm in 2 ml standard tubes. CKMix is designed for tissue homogenization like brain, liver, kidney, ear, lung, heart, spleen, tail, artery, arabidopsis, leaves, tumor, intestine, adipose tissue and more. Sample size range from 20 mg to 200 mg tissue or 200 µl to 1.6 ml culture.
Catalog No. NC1700796
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.