Learn More
Medchemexpress LLC (2R,3S,4R)-2-(hydroxymethyl)tetrahydrothiophene-3,4-diol | 190315-43-6 | MFCD27921043 | 98.7% | 150.20 g/mol | C5H10O3S | 25 MG

Supplier: Medchemexpress LLC 25MGHYW048245
Chiral hydroxymethyl tetrahydrothiophene diol supplied for research use as a 25 mg sample with high purity and supporting analytical documentation. The compound is provided as an oil for in vitro and analytical applications; COA, HNMR, and HPLC data are available.
- Chiral sulfur-containing diol.
- High purity (98.7%).
- Molecular weight 150.20 g/mol.
- Molecular formula C5H10O3S.
- Appearance: light brown oil.
- Storage: -20°C for the pure form; -80°C in solvent for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.