Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2R,5R)-1-(2-[(2R,5R)-2,5-Dimethylphospholan-1-yl]phenyl)-2,5-dimethylphospholane 1-oxide, 97+%

Catalog No. AAH6083403
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6083403 1 g
AAH60834MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6083403 Supplier Thermo Scientific Chemicals Supplier No. H6083403
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 638132-66-8 | C18H28OP2 | 322.369 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638132-66-8
Molecular Formula C18H28OP2
Molecular Weight (g/mol) 322.369
MDL Number MFCD09750450
InChI Key ORZVYNTUPREFTI-KLHDSHLOSA-N
Synonym r,r-me-bozphos, 1-2r,5r-2,5-dimethylphospholanyl-2-2r,5r-2,5-dimethylphospholanyl-1-oxide benzene, 1,2-bis 2r,5r-2,5-dimethylphospholano, 1,2-bis 2r,5r-2,5-dimethylphospholano benzene monooxide, 1,2-bis 2r,5r-2,5-dimethylphospholano benzene monooxide, kanata purity, 1,1'-1,2-phenylene bis 2r-2alpha,5beta-dimethyltetrahydro-1h-phosphole-1-one, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane 1-oxide, 97+%, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethyl-1??-phospholan-1-one, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane 1-oxide, 1-2-2alpha,5beta-dimethyltetrahydro-1h-phosphole-1-yl phenyl-2alpha,5beta-dimethyltetrahydro-1h-phosphole 1-oxide
PubChem CID 11209472
IUPAC Name (2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1$l^{5}-phospholane 1-oxide
SMILES CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C

Specifications

Melting Point 116°C to 122°C
Quantity 1 g
Sensitivity Air Sensitive
Optical Rotation −171° (c=1 in Chloroform)
Formula Weight 322.36
Percent Purity ≥97%
Chemical Name or Material (2R,5R)-1-(2-[(2R,5R)-2,5-Dimethylphospholan-1-yl]phenyl)-2,5-dimethylphospholane 1-oxide
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.