Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam (2S)-2-Hydroxyglutaric Acid, Alpha-Ketoglutarate-Dependent Dioxygenase Inhibitor, >95% Purity, 25 mg
Description
- Molecular weight: 148.11 g/mol
- Purity: >95%
- Mechanism of action: Competitively inhibits α-ketoglutarate-dependent dioxygenases, including lysine demethylases and DNA hydroxylases
- Classification: α-hydroxy acid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | α-Ketoglutarate-Dependent Dioxygenases, Lysine Demethylases, DNA Hydroxylases |
| Substrate Properties | Chemical Substrate |
| Quantity | 25 mg |
| For Use With (Application) | Inhibition of alpha-ketoglutarate-dependent dioxygenases, lysine demethylase inhibition, DNA hydroxylase inhibition |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?