Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2S,3S)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one 96.0+%, TCI America™
SDP

Catalog No. D366125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D366125G 25 g
D36615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D366125G Supplier TCI America Supplier No. D366125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42399-49-5
Molecular Formula C16H15NO3S
Molecular Weight (g/mol) 301.36
MDL Number MFCD01321316
InChI Key LHBHZALHFIQJGJ-CABCVRRESA-N
Synonym azepin-4-one, 2s,3s-+-2,3-dihydro-3-hydroxy-2-4-methoxyphenyl-1,5-benzothiazepin-4 5h-one, unii-6rn4n3305m, 2s-cis-+-2,3-dihydro-3-hydroxy-2-4-methoxyphenyl-1,5-benzothiazepin-4 5h-one, 2s,3s-3-hydroxy-2-4-methoxyphenyl-2,3-dihydro-1,5-benzothiazepin-4 5h-one, 2s,3s-3-hydroxy-2-4-methoxyphenyl-3,5-dihydro-2h-1,5-benzothiazepin-4-one, 2s-cis-2,3-dihydro-3-hydroxy-2-4-methoxyphenyl-1,5-benzothiazepin-4 5h-one, 3-hydroxy-2-4-methoxyphenyl-2,3-dihydro-1,5-benzothiazepin-4 5h-one, 2s,3s-2,3-dihydro-3-hydroxy-2-4-methoxyphenyl-1,5-benzothiazepin-4 5h-one, diltiazem impurity e
PubChem CID 2733673
IUPAC Name (2S,3S)-3-hydroxy-2-(4-methoxyphenyl)-3,5-dihydro-2H-1,5-benzothiazepin-4-one
SMILES COC1=CC=C(C=C1)C2C(C(=O)NC3=CC=CC=C3S2)O

Specifications

Melting Point 204°C
Color Yellow
Quantity 25 g
Formula Weight 301.36
Percent Purity ≥96.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (2S,3S)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one
EINECSNumber (5)-3876
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.