Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic Acid 96.0+%, TCI America™
SDP

Catalog No. B31771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31771G 1 g
B3177200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31771G Supplier TCI America Supplier No. B31771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 203866-14-2
Molecular Formula C10H16FNO4
Molecular Weight (g/mol) 233.239
MDL Number MFCD06796085
InChI Key YGWZXQOYEBWUTH-RQJHMYQMSA-N
Synonym n-boc-trans-4-fluoro-l-proline, 2s,4r-1-tert-butoxycarbonyl-4-fluoropyrrolidine-2-carboxylic acid, boc-trans-4-fluoro-l-proline, n-tert-butoxycarbonyl-2s,4r-4-fluoroproline, 2s,4r-1-boc-4-fluoro-2-pyrrolidinecarboxylic acid, boc-trans-4-fluoro-pro-oh, 2s,4r-1-tert-butoxycarbonyl-4-fluoro-2-pyrrolidinecarboxylic acid, 2s,4r-1-tert-butoxy carbonyl-4-fluoropyrrolidine-2-carboxylic acid, trans-4-fluoro-l-proline, n-boc protected, 2s,4r-1-tert-butoxycarbonyl-4-fluoropyrrolidine-2-carboyxlic acid
PubChem CID 12043048
IUPAC Name (2S,4R)-4-fluoro-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)O)F

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 1 g
Formula Weight 233.24
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.