Learn More
Medchemexpress LLC (2S,4S)-1-((S)-2-amino-3,3-dimethylbutanoyl)-4-hydroxy N-substituted pyrrolidine-2-carboxamide (hydrochloride) | 2380273-26-5 | 98.6% | 481.05 g/mol | C23H33ClN4O3S | 1 G

Supplier: Medchemexpress LLC HYW6743501G
Hydrochloride salt of a small-molecule research compound (CAS 2380273-26-5) supplied as a white to yellow solid for laboratory use. COA and SDS report purity 98.59% and provide storage and hazard guidance.
- Hydrochloride salt, solid form.
- Purity 98.6% (LCMS, COA).
- Molecular weight 481.05 g/mol.
- Chemical formula C23H33ClN4O3S.
- Recommended storage: -20°C; protect from moisture and light.
- Hazard statements: harmful if swallowed; causes skin and eye irritation; may cause respiratory irritation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.