Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone 97.0+%, TCI America™
SDP

Catalog No. B41381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41381G 1 g
B4138200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41381G Supplier TCI America Supplier No. B41381G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 346440-54-8
Molecular Formula C15H22N2O
Molecular Weight (g/mol) 246.35
MDL Number MFCD03426982
InChI Key SKHPYKHVYFTIOI-UHFFFAOYNA-N
Synonym (2S,5S)-(-)-5-Benzyl-2-tert-butyl-3-methyl-4-imidazolidinone
PubChem CID 10309834
IUPAC Name 5-benzyl-2-tert-butyl-3-methylimidazolidin-4-one
SMILES CN1C(NC(CC2=CC=CC=C2)C1=O)C(C)(C)C

Specifications

Melting Point 100°C
Quantity 1 g
Formula Weight 246.35
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.