Learn More
BIOSEARCH TECHNOLOGIES INC (Lucigen) 2X BHQ Probe Master Mix, Standard ROX, 10mLA versatile mix for both end-point PCR genotyping and qPCR applications.
Supplier:  BIOSEARCH TECHNOLOGIES INC KBS1040124
Encompass
2X BHQ Probe Master Mix, Standard ROX, 10mLA versatile mix for both end-point PCR genotyping and qPCR applications.
Catalog No. 50-223-1948