Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 2XYT-Broth, Capsules: Regular

Catalog No. MP113012031 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
454 g
1000 g
5000 g
Format:
Capsule
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
MP113012031 1000 g Capsule
MP113012021 454 g Capsule
MP113012041 5000 g Capsule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP113012031 Supplier MP Biomedicals™ Supplier No. 113012031
Add to Cart
Add to Cart

Optimized for the growth and maintenance of M13 phage or other filamentous ssDNA bacteriophages

Regular size capsules.

  • Contents/L: 16g tryptone, 10g yeast extract, 5g NaCl
  • Use 31 capsules/L

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified™
Content And Storage 15°C to 30°C
Format Capsule
Product Type Microbiology Media
Formulation Tryptone 16g/L, Yeast extract 10g/L, NaCl 5g/L
For Use With (Application) For growth and maintenance of M13 phage or other filamentous phage
Quantity 1000 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.