Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ 2XYT Broth (Granulated Media)

Catalog No. BP97432 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging Type:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP97432 2 kg Poly Pail
BP9743500 500 g Poly Bottle
BP97435 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP97432
  • $778.50
    $661.65 / Each of 1

    Save $116.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Used in the growth and cultivation of microorganisms in the lab and include E. coli, bacteriophage, and other bacterial strains

  • Granulated Media
  • Formulation per liter: Casein Peptone [91079-40-2] 16g, Yeast Extract [8013-01-2] 10g, Sodium Chloride [7647-14-5] 5g
  • Store at room temperature

TRUSTED_SUSTAINABILITY

Specifications

Form Solid
Media Type Dehydrated 2XYT
Product Type Microbiology Media
Formulation Casein Peptone 16g/L, Yeast extract 10g/L, Sodium Chloride 5g/L
Packaging Type Poly Pail
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.