Learn More
Chemglass Life Sciences Chemglass Cryogenic Vial, 3.0 mL, Self-Standing, Internal Thread, Sterile
SDP
Supplier:  Chemglass Life Sciences CGN4766012
Encompass_Preferred
Chemglass 3.0 mL cryogenic vials are self-standing, internally threaded storage vials designed for secure, long-term cryogenic sample storage. They are supplied sterile and prepared to be non-pyrogenic and DNase/RNase free, with clear graduations and a white marking area for sample identification.
  • 3.0 mL capacity.
  • Internal thread cap design.
  • Sterile; e-beam sterilized.
  • USP Class VI material.
  • Autoclavable.
  • Non-pyrogenic and DNase/RNase free.
  • Racked packaging: 9 x 9 per rack; 8 racks per case (648 vials per case).
  • Clear graduations and white marking area for identification.
  • Recommended for gas-phase nitrogen storage.
Catalog No. 50-273-3341