Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(1-Cyanoethyl)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. C273025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C273025G 25 g
C27305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C273025G Supplier TCI America Supplier No. C273025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5537-71-3
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00002520
InChI Key IRYIYPWRXROPSX-UHFFFAOYNA-N
Synonym 3-1-cyanoethyl benzoic acid, m-1-cyanoethyl benzoic acid, benzoic acid, 3-1-cyanoethyl, zlchem 504, acmc-1b01z, 3-1-cyanethyl benzoesaeure, 3-1-cyanoethyl benzoicacid, 3-l-cyanoethyl benzoic acid, 3-1-cyanethyl benzoic acid, meta-1-cyanoethyl benzoic acid
PubChem CID 98370
ChEBI CHEBI:85131
IUPAC Name 3-(1-cyanoethyl)benzoic acid
SMILES CC(C#N)C1=CC=CC(=C1)C(O)=O

Specifications

Melting Point 148°C
Color White
Quantity 25 g
Formula Weight 175.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(1-Cyanoethyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.