Learn More
Sigma Organic Chemistry 3- 1-Methyl-1H-benzimid | 1G | MFCD18483508
Supplier: Sigma Organic Chemistry CBR012751G

3- 1-Methyl-1H-benzimid , 1G
About this Item:
MDL #: MFCD18483508
Molecular Weight: 280.19
UNSPSC Code: 12352200
Chemical Formula: C11H19Cl2N3O
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.