Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(1-Pyrrolylmethyl)pyridine 97.0+%, TCI America™
SDP

Catalog No. P18531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18531G 1 g
P18535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18531G Supplier TCI America Supplier No. P18531G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 80866-95-1
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD00006405
InChI Key PJVWPGGKILHMKW-UHFFFAOYSA-N
PubChem CID 697607
IUPAC Name 3-[(1H-pyrrol-1-yl)methyl]pyridine
SMILES C(N1C=CC=C1)C1=CC=CN=C1

Specifications

Melting Point 60°C
Quantity 1 g
Formula Weight 158.20
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(1-Pyrrolylmethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.