Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(1H-Pyrrol-1-yl)benzaldehyde, 95+%, Thermo Scientific™

Catalog No. CC21904DA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC21904DA 1 g
CC21904CB 250 mg
CC21904EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC21904DA Supplier Thermo Scientific Chemicals Supplier No. CC21904DA
Add to Cart

Chemical Identifiers

CAS 129747-77-9
Molecular Formula C11H9NO
Molecular Weight (g/mol) 171.20
MDL Number MFCD03086140
InChI Key PALTUANHIBXQMX-UHFFFAOYSA-N
PubChem CID 2776528
IUPAC Name 3-pyrrol-1-ylbenzaldehyde
SMILES O=CC1=CC(=CC=C1)N1C=CC=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-1h-pyrrol-1-yl benzaldehyde, 3-pyrrol-1-yl-benzaldehyde, 3-pyrrol-1-yl benzaldehyde, benzaldehyde,3-1h-pyrrol-1-yl, benzaldehyde, 3-1h-pyrrol-1-yl, acmc-20airb, 3-pyrrolylbenzaldehyde, 3-1-pyrrolyl benzaldehyde
Formula Weight 171.2
Percent Purity 95%
Chemical Name or Material 3-(1H-Pyrrol-1-yl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.