Learn More
Medchemexpress LLC 3-[(2,4-dimethylphenyl)carbamoyl]naphthalen-2-yl dihydrogen phosphate | 1596-56-1 | MFCD00042711 | 99.5% | 371.32 | C19H18NO5P | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10406725MG
Encompass_Preferred
Naphthol AS-MX phosphate is a small-molecule inhibitor of the CREB-CBP transcription factor complex supplied as a high-purity solid for biochemical and cell-based research. It is used to probe CREB/CBP-mediated transcriptional regulation and related signaling pathways and is suitable for enzymatic assays and cellular activity studies.
  • High HPLC purity: 99.5%.
  • Solid powder form for storage and formulation flexibility.
  • Pack sizes suitable for research use, including 25 MG to 500 MG.
  • Recommended storage: 4°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Multiple authoritative identifiers available, including CAS, MFCD, and molecular formula.
Catalog No. 50-002-43465