Learn More
Sigma Organic Chemistry 3-2-4-Fluorophenyl amin 250MG
Supplier: Sigma Organic Chemistry OTV000547250MG

3-2-4-Fluorophenyl amin , 250mg
About this item:
MDL #: MFCD06009634
Chemical Formula: C16H14FNO3S
Molecular Weight: 319.35
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.