Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Aminoethyl)-5-bromoindole, 97%

Catalog No. AAH6365303
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6365303 1 g
AAH63653MD 250 mg
AAH6365306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6365303 Supplier Thermo Scientific Chemicals Supplier No. H6365303

CAS: 3610-42-2 | C10H11BrN2 | 239.116 g/mol

N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3610-42-2
Molecular Formula C10H11BrN2
Molecular Weight (g/mol) 239.116
MDL Number MFCD00130169
InChI Key CGHUQJRRADEHTQ-UHFFFAOYSA-N
PubChem CID 77158
IUPAC Name 2-(5-bromo-1H-indol-3-yl)ethanamine
SMILES C1=CC2=C(C=C1Br)C(=CN2)CCN

Specifications

Quantity 1 g
Synonym 5-bromotryptamine, 2-5-bromo-1h-indol-3-yl ethanamine, 5-bromo-1h-indole-3-ethylamine, 1h-indole-3-ethanamine,5-bromo, 2-5-bromo-1h-indol-3-yl ethan-1-amine, 1h-indole-3-ethanamine, 5-bromo, 2-5-bromo-1h-indol-3-yl-ethylamine, 5-bromo-tryptamine, 5-br-t, 3-2-aminoethyl-5-bromoindole
Formula Weight 239.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-(2-Aminoethyl)-5-bromoindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-779-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.