Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Aminoethyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. A133625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A133625G 25 g
A13365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A133625G Supplier TCI America Supplier No. A133625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20173-24-4
Molecular Formula C7H11N2
Molecular Weight (g/mol) 123.18
MDL Number MFCD00191604
InChI Key NAHHNSMHYCLMON-UHFFFAOYSA-O
Synonym 3-2-aminoethyl pyridine, 2-pyridin-3-yl ethanamine, 3-aminoethylpyridine, 3-pyridineethanamine, 2-pyridin-3-yl ethan-1-amine, 2-3-pyridyl ethylamine, 3-pyridineethaneamine, 3-pyridine ethanamine, 2-pyridin-3-yl-ethylamine, 2-3-pyridinyl ethanamine
PubChem CID 854051
IUPAC Name 2-(pyridin-3-yl)ethan-1-aminium
SMILES [NH3+]CCC1=CC=CN=C1

Specifications

Color Yellow
Boiling Point 128°C
Quantity 25 g
UN Number 2735
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(2-Aminoethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.