Learn More
Sigma Organic Chemistry 3-2-bromoacetyl benzoni | 500mg | 50916-55-7 | MFCD00833267
Supplier: Sigma Organic Chemistry CDS013717500MG

3-2-bromoacetyl benzoni , 500mg
About this Item:
CAS #: 50916-55-7
MDL #: MFCD00833267
Molecular Weight: 224.05
UNSPSC Code: 12352200
Chemical Formula: C9H6BrNO
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.