Learn More
Sigma Organic Chemistry 3-2-Hydroxyethoxy benza | 250mg | 60345-97-3 | MFCD00191448
Supplier: Sigma Organic Chemistry CDS000173250MG

3-2-Hydroxyethoxy benza , 250mg
About this Item:
CAS #: 60345-97-3
MDL #: MFCD00191448
Molecular Weight: 166.17
UNSPSC Code: 12352200
Chemical Formula: C9H10O3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.